2-(Aminomethyl)-1H-indole methanesulphonate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1H-Indol-2-yl)methylamine methanesulphonate, 2-(Aminomethyl)-1H-indole mesylate
OR5660-250MG
OR5660-1G
APOSOR5660-250MGEA
372
EUR
APOSOR5660-250MG
APOSOR5660-1G
2-(Aminomethyl)-1H-indole methanesulphonate 95%
2-(Aminomethyl)-1H-indole methanesulphonate
Formel:
C₉H₁₀N₂·CH₃SO₃H Molecular Weight: 242.295 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03756929 CAS-Nummer: 1072806-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...