7-Methyl-1H-indole-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR5661-1G
APOSOR5661-1GEA 84.1 EUR
APOSOR5661-1G
7-Methyl-1H-indole-2-carboxylic acid
7-Methyl-1H-indole-2-carboxylic acid
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047168
CAS-Nummer: 18474-60-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR