4-Methyl-3-nitrobenzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR5672-1G
APOSOR5672-1GEA
21.9
EUR
APOSOR5672-1G
4-Methyl-3-nitrobenzaldehyd 98%
4-Methyl-3-nitrobenzaldehyde
Formel:
C₈H₇NO₃ Molecular Weight: 165.15 g/mol Schmelzpunkt: 44…49 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017011 CAS-Nummer: 31680-07-6 EINECS: 250-760-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...