1-Pyridin-4-yl-N-(pyridin-4-ylmethyl)methanamine 95+%

Lieferant: Apollo Scientific

Synonyme: Bis(pyridin-4-ylmethyl)amine

OR5676-250MG
APOSOR5676-250MGEA 125 EUR
APOSOR5676-250MG
1-Pyridin-4-yl-N-(pyridin-4-ylmethyl)methanamine 95+%
1-Pyridin-4-yl-N-(pyridin-4-ylmethyl)methanamine
Formel: C₁₂H₁₃N₃
Molecular Weight: 199,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00868302
CAS-Nummer: 1539-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR