3-Acetylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Acetylphenyl)metaborsäure, Dihydroxy(3-acetylphenyl)boran, 3-Acetylbenzolboronsäure

OR5678-100G OR5678-25G OR5678-5G
APOSOR5678-100GEA 232 EUR
APOSOR5678-100G APOSOR5678-25G APOSOR5678-5G
3-Acetylphenylboronsäure
3-Acetylphenylboronsäure
Formel: C₈H₉BO₃
Molecular Weight: 163.97
Schmelzpunkt: 204…208 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01074678
CAS-Nummer: 204841-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR