3-Pyrazol-1-ylbenzylamine 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (3-(1H-Pyrazol-1-yl)phenyl)methanamine, [3-(1H-Pyrazol-1-yl)phenyl]methylamine

OR5680-1G
APOSOR5680-1GEA 91.5 EUR
APOSOR5680-1G
3-Pyrazol-1-ylbenzylamine 98%
3-Pyrazol-1-ylbenzylamine
Formel: C₁₀H₁₁N₃
Molecular Weight: 173,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03449005
CAS-Nummer: 687635-04-7
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR