3-(1H-Pyrazol-1-yl)anilin

Lieferant: Apollo Scientific

Synonyme: 3-(1-Pyrazolyl)aniline

OR5682-1G
APOSOR5682-1GEA 91.5 EUR
APOSOR5682-1G
3-(1H-Pyrazol-1-yl)anilin
3-(1H-Pyrazol-1-yl)anilin
Formel: C₉H₉N₃
Molecular Weight: 159,1879 g/mol
MDL: MFCD08059554
CAS-Nummer: 184097-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR