4-Brom-N-methylbenzolsulfonamid 98%

Lieferant: Apollo Scientific
Achtung

OR5691-1G
APOSOR5691-1GEA 21.9 EUR
APOSOR5691-1G
4-Brom-N-methylbenzolsulfonamid 98%
4-Brom-N-methylbenzolsulfonamid
Formel: C₇H₈BrNO₂S
Molecular Weight: 250,1100 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00993162
CAS-Nummer: 703-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR