N-(2-Aminoethyl)-4-brombenzolsulfonamid 98%

Lieferant: Apollo Scientific

OR5696-1G
APOSOR5696-1GEA 241 EUR
APOSOR5696-1G
N-(2-Aminoethyl)-4-brombenzolsulfonamid 98%
N-(2-Aminoethyl)-4-brombenzolsulfonamid
Formel: C₈H₁₁BrN₂O₂S
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659916
CAS-Nummer: 90002-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR