4-Brom-N-tert-butylbenzolsulfonamid 98%

Lieferant: Apollo Scientific
Achtung

OR5702-5G
APOSOR5702-5GEA 294 EUR
APOSOR5702-5G
4-Brom-N-tert-butylbenzolsulfonamid 98%
4-Brom-N-tert-butylbenzolsulfonamid
Formel: C₁₀H₁₄BrNO₂S
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00458267
CAS-Nummer: 93281-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR