4-Benzyloxy-1-tert-butoxycarbonylindole-2-boronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR5711-1G
APOSOR5711-1GEA 125 EUR
APOSOR5711-1G
4-Benzyloxy-1-tert-butoxycarbonylindole-2-boronic acid 98%
4-Benzyloxy-1-tert-butoxycarbonylindole-2-boronic acid
Formel: C₂₀H₂₂BNO₅
Schmelzpunkt: 102…104 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04973739
CAS-Nummer: 850568-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR