3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: (3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenyl)boronic acid

OR5717-1G OR5717-5G
APOSOR5717-1GEA 201 EUR
APOSOR5717-1G APOSOR5717-5G
3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure 97%
3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure
Formel: C₁₀H₁₁BN₂O₃S
Schmelzpunkt: 204…240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363799
CAS-Nummer: 871333-06-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR