3-(1,2,4-Triazol-3-yl)carbamoyl)phenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR5719-1G
OR5719-250MG
APOSOR5719-1GEA
244
EUR
APOSOR5719-1G
APOSOR5719-250MG
3-(1,2,4-Triazol-3-yl)carbamoyl)phenylboronsäure 95%
3-(1,2,4-Triazol-3-yl)carbamoyl)phenylboronsäure
Formel:
C₉H₉BN₄O₃ Schmelzpunkt: 230…236 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363798 CAS-Nummer: 871333-05-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...