2'-(Methylsulfonyl)acetanilid

Lieferant: Apollo Scientific
Achtung

OR5729-1G OR5729-25G OR5729-5G
APOSOR5729-1GEA 21.9 EUR
APOSOR5729-1G APOSOR5729-25G APOSOR5729-5G
2'-(Methylsulfonyl)acetanilid
2'-(Methylsulfonyl)acetanilid
Formel: C₉H₁₁NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD01464107
CAS-Nummer: 20628-27-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR