3-(2-Naphthyl)-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
OR5742-1G
APOSOR5742-1GEA
219
EUR
APOSOR5742-1G
3-(2-Naphthyl)-1H-pyrazole
3-(2-Naphthyl)-1H-pyrazole
Formel:
C₁₃H₁₀N₂ Molecular Weight: 194,24 g/mol Schmelzpunkt: 158…159 °C Dichte: 1,302 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569444 CAS-Nummer: 150433-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...