1-Benzyl-5-methyl-1H-1,2,3-triazole-4-carbonylchloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR5756-1G
APOSOR5756-1GEA
49.3
EUR
APOSOR5756-1G
1-Benzyl-5-methyl-1H-1,2,3-triazole-4-carbonylchloride 97%
1-Benzyl-5-methyl-1H-1,2,3-triazole-4-carbonylchloride
Formel:
C₁₁H₁₀ClN₃O Molecular Weight: 235,67 g/mol Schmelzpunkt: 94…95 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07780129 CAS-Nummer: 664351-09-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...