2-Amino-2'-chlor-5-nitrobenzophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Clonazepam Related Compound B, 2-Amino-5-nitro-2’-chlorobenzophenone, 2-Amino-2'-chloro-5-nitrobenzophenone, (2-Amino-5-nitrophenyl)(2-chlorophenyl)methanone
OR5767-5G
APOSOR5767-5GEA
21.9
EUR
APOSOR5767-5G
2-Amino-2'-chlor-5-nitrobenzophenon
2-Amino-2'-chlor-5-nitrobenzophenon
Formel:
C₁₃H₉ClN₂O₃ Molecular Weight: 276,68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792453 CAS-Nummer: 2011-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...