3-Chloro-4-(N-isopropylcarbamoyl)benzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR5776-1G
APOSOR5776-1GEA 277 EUR
APOSOR5776-1G
3-Chloro-4-(N-isopropylcarbamoyl)benzeneboronic acid 98%
3-Chloro-4-(N-isopropylcarbamoyl)benzeneboronic acid
Formel: C₁₀H₁₃BClNO₃
Molecular Weight: 241,479 g/mol
Schmelzpunkt: 184…190 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07363768
CAS-Nummer: 850589-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR