4-Methoxy-6-methyl-1,3,5-triazin-2-amin 97%

Lieferant: Apollo Scientific
Achtung

OR5791-100G
APOSOR5791-100GEA 69.8 EUR
APOSOR5791-100G
4-Methoxy-6-methyl-1,3,5-triazin-2-amin 97%
2-Amino-4-methoxy-6-methyl-1,3,5-triazine
Formel: C₅H₈N₄O
Molecular Weight: 140,1400 g/mol
Schmelzpunkt: 258…261 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052764
CAS-Nummer: 1668-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR