4-Amino-3-hydroxypyridine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR5796-250MG
OR5796-1G
APOSOR5796-250MGEA
87
EUR
APOSOR5796-250MG
APOSOR5796-1G
4-Amino-3-hydroxypyridine hydrochloride
Formel:
C₅H₆N₂O·ClH Schmelzpunkt: 222…223 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08729308 CAS-Nummer: 1206679-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...