6-(tert-Butyl)-2-(pyridin-2-yl)pyrimidin-4-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[4-(tert-Butyl)-6-hydroxypyrimidin-2-yl]pyridine, 4-(tert-Butyl)-6-hydroxy-2-(pyridin-2-yl)pyrimidine

OR5800-100MG
APOSOR5800-100MGEA 65.4 EUR
APOSOR5800-100MG
6-(tert-Butyl)-2-(pyridin-2-yl)pyrimidin-4-ol
6-(tert-Butyl)-2-(pyridin-2-yl)pyrimidin-4-ol
Formel: C₁₃H₁₅N₃O
Molecular Weight: 229,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00973775
CAS-Nummer: 874606-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR