Isochinolin-1-carbonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Isoquinolinecarbonitrile

OR5801-1G
APOSOR5801-1GEA 43.5 EUR
APOSOR5801-1G
Isochinolin-1-carbonitril 98%
Isochinolin-1-carbonitril
Formel: C₁₀H₆N₂
Molecular Weight: 154,17 g/mol
Schmelzpunkt: 81…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134166
CAS-Nummer: 1198-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR