3-Chlor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure 95%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-4-(1-pyrrolidinylcarbonyl)benzeneboronic acid, 3-Chloro-4-(pyrrolidinyl-1-carbonyl)phenylboronic acid

OR5813-1G
APOSOR5813-1GEA 302 EUR
APOSOR5813-1G
3-Chlor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure 95%
3-Chlor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BClNO₃
Schmelzpunkt: 152…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363776
CAS-Nummer: 850589-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR