3-Chlor-4-(piperidin-1-carbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-4-(1-piperidinylcarbonyl)benzeneboronic acid, (3-Chloro-4-(piperidine-1-carbonyl)phenyl)boronic acid

OR5815-1G
APOSOR5815-1GEA 302 EUR
APOSOR5815-1G
3-Chlor-4-(piperidin-1-carbonyl)phenylboronsäure 98%
3-Chlor-4-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BClNO₃
Molecular Weight: 267,52 g/mol
Schmelzpunkt: 160…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363775
CAS-Nummer: 850589-50-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR