2-Amino-4-methyl-3-nitropyridin 98%
Lieferant: Apollo Scientific
Synonyme:
4-Methyl-3-nitropyridin-2-amin, 4-Methyl-3-nitro-2-pyridylamin, 2-Amino-3-nitro-4-picolin
OR5821-100G
APOSOR5821-100GEA
81.2
EUR
APOSOR5821-100G
2-Amino-4-methyl-3-nitropyridin 98%
2-Amino-4-methyl-3-nitropyridin
Formel:
C₆H₇N₃O₂ Molecular Weight: 153,14 g/mol Schmelzpunkt: 136…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006315 CAS-Nummer: 6635-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...