3-Cyano-4-methoxy-2-pyridone 98+%
Lieferant: Apollo Scientific
|
Achtung
|
OR5829-25G
OR5829-5G
APOSOR5829-25GEA
29
EUR
APOSOR5829-25G
APOSOR5829-5G
3-Cyano-4-methoxy-2-pyridone 98+%
3-Cyano-4-methoxy-2-pyridone
Formel:
C₇H₆N₂O₂ Schmelzpunkt: 275…276 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08059559 CAS-Nummer: 21642-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...