3-Amino-2-methoxypyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR5833-5G
OR5833-1G
OR5833-25G
APOSOR5833-5GEA
24.8
EUR
APOSOR5833-5G
APOSOR5833-1G
APOSOR5833-25G
3-Amino-2-methoxypyridin 98%
3-Amino-2-methoxypyridine
Formel:
C₆H₈N₂O Schmelzpunkt: 67…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00833386 CAS-Nummer: 20265-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...