2-Acetamido-2-deoxy-4,6-O-isopropylidene-D-glucopyranose
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,6-O-Isopropylidene-GlcNAc, 2-(Acetylamino)-2-deoxy-4,6-O-isopropylidene-D-glucopyranose, 2-Acetamido-2-deoxy-4,6-O-(1-methylethylidene)-D-glucopyranose, 2-(Acetylamino)-2-deoxy-4,6-O-(1-methylethylidene)-D-glucopyranose
OR5835T-250MG
APOSOR5835T-250MGEA
122
EUR
APOSOR5835T-250MG
2-Acetamido-2-deoxy-4,6-O-isopropylidene-D-glucopyranose
2-Acetamido-2-deoxy-4,6-O-isopropylidene-D-glucopyranose
A useful synthetic intermediate for carbohydrate chemistry.
Formel:
C₁₁H₁₉NO₆ Molecular Weight: 261,27 g/mol Schmelzpunkt: 184…186 °C (Zers.) Lagertemperatur: Gefrierschrank |
MDL:
MFCD09260464 CAS-Nummer: 50605-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...