Phenoxathiin-4-boronic acid 95%

Lieferant: Apollo Scientific

Synonyme: Phenoxathiin-4-ylboronic acid

OR5866-1G
APOSOR5866-1GEA 191 EUR
APOSOR5866-1G
Phenoxathiin-4-boronic acid 95%
Phenoxathiin-4-boronic acid
Formel: C₁₂H₉BO₃S
Molecular Weight: 244.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01605731
CAS-Nummer: 100124-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR