(1S,2S)-2-Amino-1-(4-nitrophenyl)-1,3-propandiol 98%

Lieferant: Apollo Scientific

Synonyme: L-(+)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, (1S,2S)-(+)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol

OR5869-500G
APOSOR5869-500GEA 102 EUR
APOSOR5869-500G
(1S,2S)-2-Amino-1-(4-nitrophenyl)-1,3-propandiol 98%
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol
Formel: C₉H₁₂N₂O₄
Molecular Weight: 212.2026 g/mol
Schmelzpunkt: 163…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007359
CAS-Nummer: 2964-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR