7-Hydroxy-4-(pyridin-3-yl)coumarin 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR5886-1G
APOSOR5886-1GEA
63.8
EUR
APOSOR5886-1G
7-Hydroxy-4-(pyridin-3-yl)coumarin 95+%
7-Hydroxy-4-(pyridin-3-yl)coumarin
Formel:
C₁₄H₉NO₃ Schmelzpunkt: >230 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01941325 CAS-Nummer: 109512-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...