(1-Methylpiperidin-4-yl)methanamine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-methyl-piperidin-4-methylamine, (1-Methyl-4-piperidinyl)methanamine, (1-methylpiperidin-4-yl)methanamine
OR5892-1G
OR5892-5G
APOSOR5892-1GEA
21.9
EUR
APOSOR5892-1G
APOSOR5892-5G
(1-Methylpiperidin-4-yl)methanamine
(1-Methylpiperidin-4-yl)methanamine
Formel:
C₇H₁₆N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD05022430 CAS-Nummer: 7149-42-0 EINECS: 230-477-2 UN: 2735 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...