3-(Dimethylamino)piperidine 98+%
Lieferant: Apollo Scientific
|
Achtung
|
OR5899-1G
OR5899-250MG
OR5899-5G
OR5899-100MG
APOSOR5899-1GEA
0
EUR
APOSOR5899-1G
APOSOR5899-250MG
APOSOR5899-5G
APOSOR5899-100MG
3-(Dimethylamino)piperidine 98+%
3-(Dimethylamino)piperidine
Formel:
C₇H₁₆N₂ Siedepunkt: 58 °C (6,75 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD06796405 CAS-Nummer: 50534-49-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...