Methyl-3,4,5-trihydroxybenzoat 98%
Lieferant: Apollo Scientific
Synonyme:
3,4,5-Trihydroxybenzoesäuremethylester, Methylgallat, Gallussäuremethylester
OR5903-100G
APOSOR5903-100GEA
53.8
EUR
APOSOR5903-100G
Methyl-3,4,5-trihydroxybenzoat 98%
Methyl-3,4,5-trihydroxybenzoat
Formel:
C₈H₈O₅ Molecular Weight: 184,15 g/mol Schmelzpunkt: 201…203 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002194 CAS-Nummer: 99-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...