4-Methylbenzamidoxim 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Hydroxy-4-methylbenzimidamide
OR5904-1G
OR5904-25G
OR5904-5G
APOSOR5904-1GEA
36.4
EUR
APOSOR5904-1G
APOSOR5904-25G
APOSOR5904-5G
4-Methylbenzamidoxim 97%
4-Methylbenzamidoxim
Formel:
C₈H₁₀N₂O Schmelzpunkt: 140…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019952 CAS-Nummer: 19227-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...