(+/-)-3-(Boc-aminomethyl)piperidine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR5910-5G
OR5910-1G
OR5910-25G
APOSOR5910-5GEA
49.3
EUR
APOSOR5910-5G
APOSOR5910-1G
APOSOR5910-25G
(+/-)-3-(Boc-aminomethyl)piperidine 97%
(+/-)-3-(Boc-aminomethyl)piperidine
Formel:
C₁₁H₂₂N₂O₂ Schmelzpunkt: 60…70 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01632454 CAS-Nummer: 142643-29-6 UN: 3259 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...