2-(3-Bromphenyl)-5-methyl-1,3,4-oxadiazol
Lieferant: Apollo Scientific
|
Achtung
|
OR59322-1G
OR59322-5G
APOSOR59322-1GEA
125
EUR
APOSOR59322-1G
APOSOR59322-5G
2-(3-Bromphenyl)-5-methyl-1,3,4-oxadiazol
2-(3-Bromophenyl)-5-methyl-1,3,4-oxadiazole
Formel:
C₉H₇BrN₂O Molecular Weight: 239.0700 g/mol Schmelzpunkt: 79…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08691468 CAS-Nummer: 41491-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...