2-(3-Bromphenyl)-5-propyl-1,3,4-oxadiazol
Lieferant: Apollo Scientific
OR59323-1G
APOSOR59323-1GEA
205
EUR
APOSOR59323-1G
2-(3-Bromphenyl)-5-propyl-1,3,4-oxadiazol
2-(3-Bromophenyl)-5-propyl-1,3,4-oxadiazole
Formel:
C₁₁H₁₁BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878393 CAS-Nummer: 957065-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...