2-(3-Bromphenyl)-5-tert-butyl-1,3,4-oxadiazol
Lieferant: Apollo Scientific
OR59325-1G
APOSOR59325-1GEA
205
EUR
APOSOR59325-1G
2-(3-Bromphenyl)-5-tert-butyl-1,3,4-oxadiazol
2-(3-Bromphenyl)-5-tert-butyl-1,3,4-oxadiazol
Formel:
C₁₂H₁₃BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878391 CAS-Nummer: 957065-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...