1-(2,3-Dimethylphenyl)pyrrolidin

Lieferant: Apollo Scientific

OR59328-1G
APOSOR59328-1GEA 176 EUR
APOSOR59328-1G
1-(2,3-Dimethylphenyl)pyrrolidin
1-(2,3-Dimethylphenyl)pyrrolidin
Formel: C₁₂H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD09878389
CAS-Nummer: 957065-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR