7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäureethylester

Lieferant: Apollo Scientific

Synonyme: Ethyl 7-chloro-4-hydroxy-8-methylquinoline-3-carboxylate

OR59329-1G
APOSOR59329-1GEA 125 EUR
APOSOR59329-1G
7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäureethylester
7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäureethylester
Formel: C₁₃H₁₂ClNO₃
Molecular Weight: 265,6923 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173377
CAS-Nummer: 5350-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR