2-(3-Bromphenyl)-5-butyl-1,3,4-oxadiazol

Lieferant: Apollo Scientific

OR59331-1G
APOSOR59331-1GEA 205 EUR
APOSOR59331-1G
2-(3-Bromphenyl)-5-butyl-1,3,4-oxadiazol
2-(3-Bromophenyl)-5-butyl-1,3,4-oxadiazole
Formel: C₁₂H₁₃BrN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09878392
CAS-Nummer: 957065-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR