Ethyl 5-chloro-1,4-dihydro-8-methyl-4-oxoquinoline-3-carboxylate
Lieferant: Apollo Scientific
Synonyme:
Ethyl 5-chloro-4-hydroxy-8-methylquinoline-3-carboxylate
OR59338-1G
APOSOR59338-1GEA
176
EUR
APOSOR59338-1G
Ethyl 5-chloro-1,4-dihydro-8-methyl-4-oxoquinoline-3-carboxylate
Ethyl 5-chloro-1,4-dihydro-8-methyl-4-oxoquinoline-3-carboxylate
Formel:
C₁₃H₁₂ClNO₃ Molecular Weight: 265,692 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06009651 CAS-Nummer: 351893-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...