2-Chlor-6-(methylamino)pyrazin

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-N-methylpyrazin-2-amine

OR59340-500MG
APOSOR59340-500MGEA 14.7 EUR
APOSOR59340-500MG
2-Chlor-6-(methylamino)pyrazin
2-Chloro-6-(methylamino)pyrazine
Formel: C₅H₆ClN₃
Molecular Weight: 143,5700 g/mol
Schmelzpunkt: 80…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09878383
CAS-Nummer: 848366-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR