4-(2-Aminophenoxy)benzeneboronic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

OR59345-1G OR59345-250MG
APOSOR59345-1GEA 343 EUR
APOSOR59345-1G APOSOR59345-250MG
4-(2-Aminophenoxy)benzeneboronic acid hydrochloride
4-(2-Aminophenoxy)benzeneboronic acid hydrochloride
Formel: C₁₂H₁₂BNO₃·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD09878349
CAS-Nummer: 957063-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR