4-Brom-N-(4-methoxybenzyl)benzolsulfonamid
Lieferant: Apollo Scientific
OR59350-1G
APOSOR59350-1GEA
161
EUR
APOSOR59350-1G
4-Brom-N-(4-methoxybenzyl)benzolsulfonamid
4-Brom-N-(4-methoxybenzyl)benzolsulfonamid
Formel:
C₁₄H₁₄BrNO₃S Molecular Weight: 356,2349 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01359619 CAS-Nummer: 329939-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...