4-(2-Methoxy-2-oxoethoxy)benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(2-Methoxy-2-oxoethoxy)phenyl)boronic acid, Methyl 2-(4-boronophenoxy)acetate

OR59353-1G
APOSOR59353-1GEA 199 EUR
APOSOR59353-1G
4-(2-Methoxy-2-oxoethoxy)benzeneboronic acid
4-(2-Methoxy-2-oxoethoxy)benzeneboronic acid
Formel: C₉H₁₁BO₅
Molecular Weight: 209.992 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878347
CAS-Nummer: 957063-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR