3',5'-Dibromacetanilid

Lieferant: Apollo Scientific

OR59355-1G
APOSOR59355-1GEA 176 EUR
APOSOR59355-1G
3',5'-Dibromacetanilid
3',5'-Dibromacetanilid
Formel: C₈H₇Br₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09878317
CAS-Nummer: 119430-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR