4-Brom-N-(4-methoxybenzyl)-2-nitroanilin

Lieferant: Apollo Scientific

OR59357-1G
APOSOR59357-1GEA 102 EUR
APOSOR59357-1G
4-Brom-N-(4-methoxybenzyl)-2-nitroanilin
4-Brom-N-(4-methoxybenzyl)-2-nitroanilin
Formel: C₁₄H₁₃BrN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09878316
CAS-Nummer: 957062-86-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR