N-p-Tolyl-4-boronobenzolsulfonamid
Lieferant: Apollo Scientific
Synonyme:
4-[(4-Methylphenyl)sulphamoyl]benzeneboronic acid
OR59358-1G
APOSOR59358-1GEA
176
EUR
APOSOR59358-1G
N-p-Tolyl-4-boronobenzolsulfonamid
N-p-Tolyl-4-boronobenzolsulfonamid
Formel:
C₁₃H₁₄BNO₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878329 CAS-Nummer: 957062-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...